Learn More
Norovirus group II Mouse anti-Virus, Clone: HM024, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13625
Catalog No. 89-033-588
Add to cart
Description
Description
Mouse monoclonal antibody raised against norovirus group II.
  • High specificity
  • High reactivity
Specifications
Specifications
Norovirus group II
HM024
Mouse monoclonal antibody raised against norovirus group II.
In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with group II capsid protein.
ELISA, Lateral Flow, Western Blot
Unconjugated
ELISA Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG2b
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)