Learn More
Honeywell™ North™ 4000 Series Full Facepiece Respirators
The 4000 Full Facepiece respirator is an industry classic in black and with welding attachment options.
Manufacturer:  Honeywell™ 410010
 View more versions of this product
Catalog No. 18-253
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Durable silicone facepiece has wide sealing flanges for comfort and seal integrity with a minimum of headstrap pressure.
  • Single-curve polycarbonate lens provides wearer with an unobstructed wide-angle view.
  • Inside surface features an antifog coating effective in temperatures as low as -31.6°C (-25°F), providing unimpaired vision in even the coldest climates.
  • Offset browguard and special surface coating protect lens from scratches.
  • Four-point cradle headstrap maintains a secure fit with comfortable tension.
  • An advanced speaking diaphragm allows clear communication with less annoying resonance.
With minor conversions, the full facepiece can be used with other Survivair respirators. Meets NIOSH 42CFR84.
Specifications
Specifications
Small