Learn More
North 5500 Series Convenience Pack Respiratorsn
Manufacturer:  Honeywell Safety Products 5503M
 View more versions of this product
Catalog No. 19-999-218
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Organic Vapor, Acid Gas Cartridge
Black
Medium half mask air purifying respirator and organic vapor or acid gas cartridge
0.9 lb.
Respirator Convenience Pack
NIOSH
Exposure to organic vapors such as mek and isocyanates plus acid gassuch as chlorine, sulfur dioxide and hydrogen fluoride
Elastomer
0.4kg
Medium