Learn More
Honeywell™ North™ A400 Series Safety Glasses
Sporty, lightweight safety glasses with one-piece wraparound lens
Manufacturer:  Honeywell™ A400
 View more versions of this product
Catalog No. 19-053-701A
Add to cart
Description
Description
  • Single lens and frame made of impact-resistant polycarbonate
  • Comfortable, wraparound design provides excellent peripheral vision protection without the need for sideshields
  • Feature antifog coating
Compliance
Meet ANSI™ Z87.1-2003 and CSA Z94.3 standards.
Specifications
Specifications
Meet ANSI Z87.1-2003 and CSA Z94.3 standards.
Clear
Scratch-resistant
Safety
Clear