Learn More
Honeywell The Peak A79 Hard Hats
Versatile, comfortable design provides dependable, all-day head protection
Manufacturer:  Honeywell A79090000
 View more versions of this product
Catalog No. 19-042-022
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Meets ANSI Z8.1-2003; Type I; Class E.
Grey
Pin-Lock