Learn More
Honeywell™ North Adaptec™ Safety Glasses
Catalog Number 17986714
Click to view available options
Description:
Narrow Size, Dark Blue Frame, Clear Lens
Narrow Size, Dark Blue Frame, Smoke Lens
Narrow Size, Replacement Lens, Clear Lens
Narrow Size, Translucent-Black Frame, Amber Lens
Narrow Size, Translucent-Black Frame, I/O Mirror Lens
Narrow Size, Translucent-Black Frame, IR Shade 5.0 Lens
Narrow Size, Translucent-Black Frame, Smoke Lens
Regualr Size, Replacement Lens, Clear Lens
Regular Size, Black Frame, Amber Lens
Regular Size, Dark Blue Frame, Clear Lens
Regular Size, Translucent-Black Frame, Amber Lens
Regular Size, Translucent-Black Frame, Clear Lens
Regular Size, Translucent-Black Frame, I/O Mirror Lens
Regular Size, Translucent-Black Frame, IR Shade 3.0 Lens
Regular Size, Translucent-Black Frame, IR Shade 5.0 Lens
Regular Size, Translucent-Black Frame, Smoke Lens
Wide Size, Black Frame, Amber Lens
Wide Size, Black Frame, Clear Lens
Wide Size, Black Frame, Smoke Lens
Wide Size, Dark Blue Frame, Clear Lens
Wide Size, Dark Blue Frame, Smoke Lens
Wide Size, Replacement Lens, Clear Lens
Wide Size, Translucent-Black Frame, Amber Lens
Wide Size, Translucent-Black Frame, Clear Lens
Wide Size, Translucent-Black Frame, IR Shade 3.0 Lens
Wide Size, Translucent-Black Frame, IR Shade 5.0 Lens
Wide Size, Translucent-Black Frame, Smoke Lens
This item is not returnable. View return policy
This item is not returnable. View return policy
Honeywell™ North Adaptec™ Safety Glasses deliver excellent protection, superior comfort, customized adjustability, premium optics and stylish look.
  • Adjustable nose bridge centers eyes to reduce fatigue
  • Self-adjusting nose pads provde greater comfort, contact and stability
  • Vented cushioned frame allows optimum air circulation to reduce fogging
  • Temple hinge ratchets to seven positions — 30° of adjustment— to angle lens angle for optimal eye protection
  • Temple design adjusts tension for personal comfort
  • 99.9% UV protection
  • Replaceable lenses
  • Adjustable temples for comfort
  • Available in many lens tints, including SCT
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Certifications/Compliance Meets ANSI Z87.1-2010, CSA Z94.3
Lens Coating Anti-static, Scratch-resistant
Description Regular Size, Translucent-Black Frame, Smoke Lens
Lens Tint Smoke
Product Type Safety glasses
Frame Color Translucent Black
Frame Material Polycarbonate
Frame Type Regular