North Adhesive Fingertip Bandages

Fabric stretches with every movement

Manufacturer: Honeywell Safety Products 011360

 View more versions of this product

Catalog No. 19-310-030

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Adhesive Aggressive adhesive
    Color Beige
    For Use With (Application) For a wide range of wound care applications
    Material Woven
    Product Type Fingertip T Bandage
    Quantity 100 Pack

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit