North Adhesive Knuckle Bandages

Greater protection for wound care applications

Manufacturer: Honeywell Safety Products 010810

 View more versions of this product

Catalog No. 19-070-450

  Check Availability
Add to Cart

    Specifications

    Adhesive Aggressive adhesive
    Color Beige
    For Use With (Application) For effective first-aid treatment, wide range of wound care applications
    Material Heavy Woven
    Product Type Knuckle Bandage
    Quantity 50/Box
    Size 7/8 in. x 3 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit