North™ AK Cleanroom Rubber Gloves

Natural rubber latex cleanroom chemical-resistant gloves

Manufacturer: Honeywell Safety Products AK1815/O/10

 View more versions of this product

Catalog No. 19-067716

  Check Availability
Add to Cart

Alert

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


    Specifications

    Glove Cuff Style Beaded
    Length (Metric) 38cm
    Material Rubber
    Powdered No
    Thickness 20mil
    Color Orange
    Length (English) 15 in.
    Size 10

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit