Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Aspirin Pain Relievers

For temporary relief of minor aches and pains

$28.49

Specifications

Format Tablet
For Use With (Application) Pain relief of headaches, muscular aches and pains
Inactive Ingredients Corn Starch, Povidone, Silicon Dioxide, Stearic Acid, Croscarmellose Sodium, Hypromellose, Microcrystaline Cellulose, mineral oil, PEG, Propylene Glycol, Titanium Dioxide contains one or more of these ingredients
Storage Requirements 15° to 30°C
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
17-986-437
View Documents
Honeywell™
163250
Extra Strength Aspirin Pain Reliever 250 Pk.
Each for $28.49
Add to cart
 
17-986-422
View Documents
Honeywell™
161512
Aspirin Tablets 250 Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Buffered with three buffering agents
  • Provides all the benefits of aspirin, but is gentler on the stomach
  • Compares to Bufferin™
  • Helps reduce fever

Specifications

Specifications

Tablet
Pain relief of headaches, muscular aches and pains
Corn Starch, Povidone, Silicon Dioxide, Stearic Acid, Croscarmellose Sodium, Hypromellose, Microcrystaline Cellulose, mineral oil, PEG, Propylene Glycol, Titanium Dioxide contains one or more of these ingredients
15° to 30°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit