Learn More
Honeywell™ North™ Biohazard PPE Kit
Provides basic personal protection against a wide-range of potentially infectious microorganisms.
$324.47 - $419.00
Specifications
Product Type Biohazard response kit
For Use With (Application) For use by medical professionals, law enforcement, correctional facilities and emergency personnel in infectious control, offers basic protection for during response or cleanup activities
View More Specs
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Includes: All kit contents identical (see table) except for coverall size. Small kit contains Medium coverall, medium kit contains Large coverall, large kit contains X-Large coverall.
Products 2
Catalog Number Mfr. No. Includes Size Price Quantity & Availability  
Catalog Number Mfr. No. Includes Size Price Quantity & Availability  
19-165-242
View Documents
Honeywell™
130023L
1 Zip lock bag, 10 antiseptic wipes, 1 reusable Poly bag, 1 chip insert biohazard, 1 Tyvek™ 2XL clear coverall, 10 Sanizide Plus™ germicidal wipes, 1 2XL latex bootie, 2 biohazard bags, 2 red twist ties, 1 indirect gogg Large
Case of 5 for $324.47
Add to cart
19-165-241
View Documents
Honeywell™
130023M
1 Zip lock bag, 10 antiseptic wipes, 1 reusable Poly bag, 1 chip insert biohazard, 1 Tyvek™ large clear coverall, 10 Sanizide Plus™ germicidal wipes, 1 Large latex bootie, 2 biohazard bags, 2 red twist ties, 1 indirect Medium
Case of 5 for $419.00
Add to cart
Description
Description
Designed for use by medical professionals, law enforcement, correctional facilities and emergency personnel in infectious control, this kits offers basic protection for during response or cleanup activities.
Kit offers limited protection from biological exposures during:
  • Natural disaster response
  • Clean-up of sites exposed to influenza virus
  • Professional clean-up activities involving the Norovirus typically found in healthcare institutions, cruise ships, schools, hotels, and restaurants
Compliance
Meets World Health Organization (WHO) Protocol Recommendations for Personal Protective Equipment (PPE)
Specifications
Specifications
Biohazard response kit
For use by medical professionals, law enforcement, correctional facilities and emergency personnel in infectious control, offers basic protection for during response or cleanup activities