Learn More
Honeywell Safety Products North Bloodborne Pathogen Response Kits
Provide fast, effective cleanup of potentially infectious materials
$102.30
Specifications
Certifications/Compliance | OSHA 1910.130 |
---|---|
Includes | One 2 oz. absorbent , 1 absorbent towel, 1 antiseptic wipes, 24 x 24 in. 2 biohazard bag, with ties, 1 germicidal surface wipe, 1 gown, 2 medical gloves, pairs, 1 scraper, 1 shovel, 1 surgical mask and shield |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-130-2507
|
Honeywell Safety Products
0197404526 |
24 Unit Bloodborne Pathogen Response Kit |
Each for $102.30
|
|
Description
Include necessary supplies to clean up a spill containing bloodborne pathogens, such as blood, vomit, and other bodily fluids.
- Easy to usesimply cover spill with absorbent beads, allow to congeal, scoop up spill with shovel, and place into biohazard bag
- Packaged in sealed plastic bags
- Intended for single use
Dispose of contaminated items and waste properly in accordance with local, state, and federal regulations.
Compliance
Includes personal protective equipment necessary to protect employees in accordance with OSHA 1910.130.
Specifications
OSHA 1910.130 | |
One 2 oz. absorbent , 1 absorbent towel, 1 antiseptic wipes, 24 x 24 in. 2 biohazard bag, with ties, 1 germicidal surface wipe, 1 gown, 2 medical gloves, pairs, 1 scraper, 1 shovel, 1 surgical mask and shield |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.