North Blue 1 Half Mask Respirators

Manufacturer: Honeywell Safety Products B210010

 View more versions of this product

Catalog No. 19-068523

  Check Availability
Add to Cart

    Specifications

    Cartridge Type S Series Cartridge
    Color Blue
    For Use With (Application) Fire protection, government, manufacturing, medical, military, mining, pharmaceutical, steel and metals
    Includes Silicone skirt, speaking diaphragm, dual sealing flange with one inner and one outer flange, rear split strap, retainer clips
    Material Silicone
    Product Type Half Mask Respirator
    Size Small

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit