North Bulk First Aid Kits

Designed for large facilities

Manufacturer: Honeywell Safety Products 019701-4501F/DEL

 View all options for this product

Catalog No. 19-130-2484


This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Certifications/Compliance ANSI Z308.1-2003
    Product Type First Aid Kit
    For Use With (Application) Manufacturing settings where a wide variety of first aid items are required
    Includes Sixteen 1" x 3" Adhesive bandages, Plastic Adhesive bandage, fingertip, Cloth Adhesive bandage, extra large, Plastic 2 Adhesive tape, 1/2" x 2-1/2 yd. 6 Antiseptic wipes, 2" x 6 yd. BZK conforming bandage, 3" x 6 yd., non-sterile conforming bandage, non-s
    Material Metal
    Sufficient For 10 Persons

    • Contain economical bulk supplies that can be readily accessed when needed
    • Kits are ideal for manufacturing settings where a wide variety of first aid items are required
    • Latex-free
    • Weatherproof
    • Supplies housed in waterproof metal or plastic case

  • Description & Specifications
  • Accessories

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit