North Bulk First Aid Kits Get IT Item

Designed for large facilities

Manufacturer: Honeywell Safety Products 0197044504

 View more versions of this product

Catalog No. 19-130-2487

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ANSI Z308.1-2003
    For Use With (Application) Manufacturing settings where a wide variety of first aid items are required
    Includes Hundred 1 x 3 in. Adhesive bandage, plastic, 4 Adhesive bandage, fingertip, cloth, 2 Adhesive bandage, extra large, 3 Adhesive bandage, knuckle, cloth, Two 0.5 in. x 2.5 yd. Adhesive tape, 12 Antimicrobial towelettes, Three 2 in. x 6 yd. conforming bandage nonsterile, two 3 in. x 6 yd. conforming bandage, 2 in. Elastic bandage, Eye-Lert™, 1 oz. eyewash, 2 Eye pad, sterile, 12 First aid or burn cream with aloe, First aid guide, Forceps, pointed Plastic, Ten 2 x 2 in. Gauze pads sterile, Two 3 x 3 in. Gauze pads sterile, 2 Instant cold pack, 4 Pair Medical gloves, synthetic
    Material Plastic
    Sufficient For 50 Persons
    Product Type First Aid Kit

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit