North Bulk First Aid Kits

Designed for large facilities

Manufacturer: Honeywell Safety Products 019707-4507F/DEL

 View more versions of this product

Catalog No. 19-130-2490

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Certifications/Compliance ANSI Z308.1-2003
    For Use With (Application) Manufacturing settings where a wide variety of first aid items are required
    Material Metal
    Product Type First Aid Kit
    Sufficient For 75 Persons

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit