Learn More
Honeywell North™ Butyl Gloves
High permeation resistance to gas or water vapors
Manufacturer:  Honeywell B131/9
 View more versions of this product
Catalog No. 11-394-9C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Smooth
Hand-specific
Butyl
13mil
Black
27.9cm
Beaded/Rolled
Ketones, Esters, high permeation resistance to Gas or Water vapors
9
ANSI/ISEA, ASTM, EN, 29 CFR
11 in.