Honeywell North™ Butyl Gloves

High permeation resistance to gas or water vapors

Manufacturer: Honeywell B161/8

 View more versions of this product

Catalog No. 19-025591

  Check Availability
Add to Cart

    Specifications

    Certifications ANSI/ISEA, ASTM, EN, 29 CFR
    Exterior Finish Smooth
    Glove Cuff Style Beaded, Rolled
    Hand Hand-specific
    For Use With (Application) Ketones, Esters, high permeation resistance to Gas or Water vapors
    Material Butyl
    Size 8
    Thickness 16mil
    Color Black
    Length (English) 11 in.
    Length (Metric) 27.9cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit