Learn More
Honeywell™ North™ Butyl Gloves
High permeation resistance to gas or water vapors
Manufacturer:  Honeywell™ B324R/8
 View more versions of this product
Catalog No. 11-395-45A
Add to cart
Description
Description
For use with ketones and esters, and applications such as aircraft maintenance, automotive, chemical handling and processing and hazmat work.
  • Made from butyl synthetic rubber
  • All gloves have rolled cuffs
  • Available with smooth or rough Grip Saf™ finish for wet applications
  • Unlined wtih sanitized interior
Compliance
Meets MIL-DTL-43976D specifications requirements.
Specifications
Specifications
Safety
Beaded/Rolled
domestic preparedness, hazmat applications
8
Black
35.5 cm
Rough grip saf
Hand-specific
Butyl
32 mil
14 in.
ANSI/ISEA, ASTM, EN, 29 CFR