Learn More
Honeywell™ North™ Butyl Gloves
High permeation resistance to gas or water vapors
Manufacturer:  Honeywell™ B131R/11
 View more versions of this product
Catalog No. 19-822-611
Add to cart
Description
Description
For use with ketones and esters, and applications such as aircraft maintenance, automotive, chemical handling and processing and hazmat work.
  • Made from butyl synthetic rubber
  • All gloves have rolled cuffs
  • Available with smooth or rough Grip Saf™ finish for wet applications
  • Unlined wtih sanitized interior
Compliance
Meets MIL-DTL-43976D specifications requirements.
Specifications
Specifications
Rough grip saf
Hand-specific
Butyl
13mil
11 in.
MIL-DTL-43976D
Beaded/Rolled
Ketones, Esters, high permeation resistance to Gas or Water vapors
11
Black
27.9cm