Learn More
Honeywell North™ Butyl Gloves
High permeation resistance to gas or water vapors
Manufacturer:  Honeywell B161R/11
 View more versions of this product
Catalog No. 19-025599
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rough grip saf
Hand-specific
Butyl
16mil
Black
27.9cm
Beaded/Rolled
Ketones, Esters, high permeation resistance to Gas or Water vapors
11
ANSI/ISEA, ASTM, EN, 29 CFR
11 in.