Learn More
Honeywell™ North Conspire™ Safety Eyewear
Click to view available options
Lens Tint:
Amber
Clear
Silver Mirror
Silver, Indoor/Outdoor Mirror
TSR Gray
Frame Color:
Matte Amber
Matte Clear
Matte Gray
Lens Coating:
Scratch-resistant
Description
- Provides 99.9% UV protection
- Lightweight, 3threepiece full-wrap design
- Shaped nosepiece and flexible temples
- Dielectric with no metal parts
- Eight-base single front with dual lenses
- Available in lens tints with Hard coat and Fog-Ban™ Anti-Fog coatings
Applications:
Construction, manufacturing industries.
Compliance
Meets the ANSI™ Z87.1-2010 and and CSA Z94.3 standards.
Specifications
Lens Type | Uvextra Anti-Fog |
Lens Tint | TSR Gray |
Product Type | Safety Eyewear |
For Use With (Application) | Construction, Manufacturing |
Frame Color | Matte Gray |