Learn More
North Cartridge Filter Accessory, Seal Check/Filter Cover
Manufacturer:  Honeywell Safety Products N750027
Catalog No. 17-623-18
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • For 7506N95, 7506N99 and 7506R95 filters
  • Attaches to gas and vapor cartridges
  • For filter use only, use N750015 filter hold
Description & Specifications
Specifications
7506N95, 7506N99 and 7506R95 Filter
Seal Check/Filter Cover