Learn More
Honeywell™ North™ CFR-1™ Respirator Replacement Filters
Manufacturer:  Honeywell™ 42N95
Catalog No. 19-051-335
Add to cart
Description
Description
N95 replacement filters for reusable CFR-1 respirators offer minimum 95% efficiency filtration of solids and particulate aerosols free of oil. Suitable for applications in dusty environments including grinding, sanding, and sweeping. When used with Cat. Nos. 19-051-339 and 19-051-340, these filters are also useful for applications such as brazing, torch cutting, soldering, and metal pouring.
Order Info
Sold in packs of 20 filters.
Specifications
Specifications
NIOSH
Welding, grinding and sanding
Safety
Filter Cartridge
10 Case
Cfr-1 respirator
Particulate Filter