North CFR-1™ Reusable Particulate Respirators

Replaceable filters save money and reduce waste

Manufacturer: Honeywell Safety Products 4200L

 View more versions of this product

Catalog No. 19-051-338

  Check Availability
Add to Cart

    Specifications

    Certifications NIOSH
    Includes Single N95 filter for minimum 95% efficient filtration of solids and particulate aerosols free of oil.
    Latex Free Yes
    Suspension Type Cradle
    Product Type Dust Mask
    Quantity 1
    Disposable No
    Filter Type N95 Filter
    For Use With (Application) Grinding, sanding, sweeping, dusty environment
    Material (Faceshield) Thermoplastic Elastomer
    Size Large
    Color Blue
    Color Code White

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit