North Compact Air Powered Air-Purifying Respirators: Filter Cartridge Assembly

For Air powered air purifying respirator systems

Manufacturer: Honeywell Safety Products 4001HE

 View more versions of this product

Catalog No. 19-134-260

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Compact air powered air purifying respirator,Compact air powered air purifying respirator
    Product Type Organic Vapor Cartridge
    For Use With (Application) Locations where movement is critical or the area to cover exceeds 300 ft., best for occasional use that may not warrant the investment in air supply system
    Includes Blower and battery assembly, belt and backpad, flow meter, battery charger and carrying case, multiple alarm
    Certifications NIOSH
    Filter Type HEPA Filter
    Color Black, Magenta

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit