Learn More
Honeywell™ North™ Compact Air Powered Air-Purifying Respirators: Primair Facepiece
Replacement hood for Primair™ 101M hood assembly is a loose fitting facepiece with an under the chin, nylon collar. Size medium, 10/BX. Hoods have an assigned protection factor (APF) of 1000.
$96.77 - $337.00
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-185-017
View Documents
Honeywell™
PA102S
Small
Pack of 10 for $337.00
Add to cart
19-134-238
View Documents
Honeywell™
PA102M
Medium
Pack of 10 for $96.77
Add to cart
Description
Description
The Primair™ 100 Series includes one loose fitting head cover and several hoods that can be used with the CA200 Series PAPR and the CF1000 Series CF-SAR. The manifold directs air over lens to reduce fogging. Triple adjustment headgear for a comfortable fit.
Certifications
NIOSH approved.