North Cough and Cold Relief

Relieve cold and cough discomfort

Manufacturer: Honeywell Safety Products 210100

 View more versions of this product

Catalog No. 17-986-455

  Check Availability
Add to Cart

    Specifications

    Active Ingredients Menthol 6.1mg
    Flavor Cherry
    For Use With (Application) Provide soothing relief of scratchy, sore throats to help prevent coughs
    Format Lozenge
    Quantity 100 Pack
    Product Type Cough Drops

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit