North Emergency Response Kits

Contains chlorinated absorbent beads with 10,000ppm of stabilized chlorine

Manufacturer: Honeywell Safety Products 127003

 View more versions of this product

Catalog No. 17-206-5

  Check Availability
Add to Cart

    Specifications

    Capacity 2 gal., 7.6L
    Includes One 2 oz. vial absorbent beads, 1 poison control directory, 1 shovel, 1 towel, 1 hand wipe, 1 surface wipe, 1 biohazard bag, 1 bag tie, 2 gloves
    Capacity (English) 2 gal.
    Certifications/Compliance OSHA
    Color Red
    Capacity (Metric) 7.6L
    Product Type Emergency Response Kit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit