Learn More
Honeywell™ North™ First Aid Self-Adhesive Wrap
Self-Adherent Wraps are sold separately and are also a component of many first aid kits. Length: 5 yards. Choice of width: 2, 3 or 4 inches.
Manufacturer:  Honeywell™ 720072
 View more versions of this product
Catalog No. 19-074580
Add to cart
Description
Description
Used to secure and protect primary dressings and other devices. Also helps immobilize injuries and provide compression. Self-adherent wrap sticks to itself without the need for adhesive, pins or clips.
Specifications
Specifications
Agriculture, chemical, construction, fire protection, food services, forestry, medical, law enforcement, military, mining, municipal services, nuclear, oil and gas, pharmaceutical, steel and metals, transportation, utilities, welding, wind energy
Self Adherent Wrap
Safety
3 in. x 5 yd.