Learn More
Honeywell™ North™ Fuse™ Safety Glasses
Click to view available options
Lens Coating:
Scratch-resistant
Lens Tint:
Clear
Silver Mirror
TSR Gray
Description
Deliver exceptional coverage with both style and comfort
• Lightweight, durable, gunmetal frame
• Dual 9–base wrap-around lens design for full unobstructed field of vision
• Adjustable saddle nose pads, soft temple tips
• Durable polycarbonate lenses with scratch-resistant or Fog-Ban™ antifog coating
• Adjustable breakaway Flexicord™ included
Compliance
Meet ANSI Z87.1-2010 and CSA Z94.3 safety standards
Specifications
Lens Coating | Scratch-resistant |
Certifications/Compliance | Meet ANSI Z87+ and CSA Z94.3 safety standards |
Lens Tint | Silver Mirror |
Frame Color | Gun Metal |
Includes | Break-Away Flexicord |
Lens Material | Polycarbonate |