North Fuse™ Safety Glasses
Deliver exceptional coverage with both style and comfort
Manufacturer: Honeywell Safety Products 11150806
 View more versions of this product
Catalog No. 19-130-0843
  Check Availability
Add to Cart
    Specifications
    Frame Color Gun Metal
    Lens Coating Fog-Ban Antifog, Scratch-Resistant, Hardcoat
    Includes Break-Away Flexicord
    Lens Material Polycarbonate
    Lens Tint TSR Gray
  • Description & Specifications
Related Products
Product Content Feedback