North Gauze Pads

Sterile, 12-ply pads

Manufacturer: Honeywell Safety Products 067533

 View more versions of this product

Catalog No. 19-014-853

  Check Availability
Add to Cart

    Specifications

    Dimensions (L x W) 3 x 3 in.
    For Use With (Application) Ideal for cleaning a wound or applying medication
    Grade USP surgical
    Material Mesh
    Thickness 12 ply
    Product Type Gauze Pad
    Quantity 25 Pack
    Sterile Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit