Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Gauze Pads

Sterile, 12-ply pads

$2.87 - $28.25

Specifications

Type Gauze Pad
Sterility Sterile
For Use With (Application) Ideal for cleaning a wound or applying medication
Grade USP surgical
Material Mesh
View More Specs

Products 9
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
17-986-467
View Documents
Honeywell™
067522
2 X 2 in. 25 Pack
Pack for $5.75
Add to cart
 
17-986-471
View Documents
Honeywell™
067544
4 X 4 in. 25 Pack
Pack for $9.51
Add to cart
 
17-986-472
View Documents
Honeywell™
067644
4 X 4 in. 100 Pack
Pack for $28.25
Add to cart
 
17-986-468
View Documents
Honeywell™
067622
2 X 2 in. 100 Pack
Pack for $13.85
Add to cart
 
17-986-470
View Documents
Honeywell™
067633
3 X 3 in. 100 Pack
Pack for $19.34
Add to cart
 
19-310-035
View Documents
Honeywell™
067422
2 X 2 in. 10 Pack
Pack of 10 for $2.87
Add to cart
 
19-310-036
View Documents
Honeywell™
067444
4 X 4 in. 10 Pack
Pack of 10 for $4.60
Add to cart
 
17-986-469
View Documents
Honeywell™
067433
3 X 3 in. 10 Pack
Pack for $3.14
Add to cart
 
19-014-853
View Documents
Honeywell™
067533
3 x 3 in. 25 Pack
Pack of 25 for $6.48
Add to cart
 
Description

Description

Ideal for cleaning a wound or applying medication. Pads are made from USP surgical gauze mesh. Available in a variety of sizes.

Specifications

Specifications

Gauze Pad
Ideal for cleaning a wound or applying medication
Mesh
Sterile
USP surgical
12 ply
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit