Learn More
North General Purpose Emergency Burn Kit Products for Science Education
Contains both dry and wet dressings for minor burn emergencies.
Manufacturer:  Honeywell Safety Products 0197270014L/EMD
Catalog No. S98412
Add to cart
Includes: Two Water-Jel™ burn dressing packets (4 x 16 in. [10.2 x 40.6cm] and 4 x 4 in. [10.2 x 10.2cm]); burn sheet with safety pins; 4 x 6 in. (10.2 x 15.2cm) sterile gauze bandage; 4 oz. sterile eyewash; 1 in. x 5 yd. (2.5 x 22.9m) adhesive tape; scissors; and two pairs of latex gloves
Description
Description
Dry dressings protect against contamination while wet dressings help draw heat out of the burn.
  • Plastic kit mounts on wall and features a carrying handle for easy transport
  • Dimensions: 9.5L x 9.5W x 2.6 in.H (22 x 9 x 15cm)
Specifications
Specifications
9.5 x 9.75 x 2.6 in. (24 x 26.67 x 7cm)
2.6 in.
Two Water Jel™ burn dressing packets and burn sheet with safety pins, sterile gauze bandage, 4 oz. sterile eyewash, adhesive tape, scissors and two pairs of latex gloves
24cm
9.75 in.
Protect against contamination while wet dressings help draw heat out of the burn
7cm
9.5 in.
Emergency Burn Kit
26.67cm