North Grip N™ Hot Mill Gloves
Heavy-duty cotton gloves ideal for workers in millwork or heavy labor
Manufacturer: Honeywell Safety Products 52/7406
 View more versions of this product
Catalog No. 19-150-0982
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications USDA, FDA, 21 CFR
    Glove Cuff Style Knit
    Hand Ambidextrous
    For Use With (Application) automotive assembly, food processing, workers in millwork or heavy labor
    Material Kevlar™
    Size Universal
    Temperature (English) Max. 400°F
    Temperature (Metric) Max 204°C
    Coating Material Nitrile
    Cut Resistance Rating Level 3
  • Description & Specifications
Related Products
Product Content Feedback