Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Grip N™ Hot Mill Gloves with Double-Sided Nitrile Dot Grip

Catalog Number 19025803
Click to view available options
One Size
This item is not returnable. View return policy
This item is not returnable. View return policy

Knit cotton gloves feature nitrile dots on both sides for enhanced grip, and protection against heavy or sharp objects.

  • Knit construction conforms snugly to the hand and provides a high level of dexterity
  • Cotton lining reinforces the glove's mechanical performance, while also absorbing perspiration
  • High quality materials create an effective barrier against heat, and heavy or sharp parts
  • Double sided nitrile dot coating enhances grip capabilities
  • Ambidextrous design is cost-effective
TRUSTED_SUSTAINABILITY
Certifications/Compliance (Europe) EN 407
Coating Material Nitrile
Color Natural
Hand Ambidextrous
Material Cotton
Product Type Hot Mill Gloves
Size Universal
Temperature (English) Max. 482°F
Temperature (Metric) Max 250°C
For Use With (Application) Automotive Assembly, Food Processing, Millwork, and Heavy Labor
Glove Cuff Style Knit
Series 51/7147
Yarn Gauge 7
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit