North Grip N™ Nitrile Gloves

Nitrile N-Tread™ coating improves grip, abrasion resistance and durability

Manufacturer: Honeywell Safety Products 79/1191S

 View more versions of this product

Catalog No. 18-999-2491

  Check Availability
Add to Cart

    Specifications

    Certifications USDA, FDA, 21 CFR
    Glove Cuff Style Knit
    Hand Ambidextrous
    For Use With (Application) automotive assembly, general maintenance work, light assembly, manufacturing, small part handling
    Material Cotton, Polyester
    Size Universal, Women's
    Coating Material Seamless
    Color Cream

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit