Learn More
Honeywell™ North™ Insect and Animal Repellent Sprays
For outdoor, on-the-job protection from insects and aggressive animals
Manufacturer:  Honeywell™ 113890
 View more versions of this product
Catalog No. 19-050216
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Wasp, Bee, and Hornet Spray
·    Patented formula kills wasps, bees, spiders, and hornets on contact
·    Range of spray is 10 ft. (3m), so insecticide can safely saturate nests in electrical switch gears, motors, and other machinery
·    Dielectric, nonconducting up to 26,500V
·    15 oz. (425g) spray can
Insect Repellent
·    Protects against mosquitoes and other biting insects
·    Active ingredient: 23.75% DEET (n,n-diethyl-m-toluamide)
·    5 oz. (142g) spray can
SPRAY HALT II™ Animal Repellent
·    For animal attacks only
·    Active ingredient, 1% capsaicin (cayenne pepper)
·    Causes burning sensation in animal's eyes and nose and on skin
·    Does not inflict permanent harm
·    1.5 oz. (43g) spray can
Specifications
Specifications
Repellent Spray
147.8mL
5 oz.
Safety
For outdoor, on-the-job protection from insects and aggressive animals, protects against mosquitoes and other biting insects