Learn More
Honeywell™ North™ LGT Eyewear
Click to view available options
Nd:Yag, Holmium, Erbium and Carbon dioxide, fiber optics diodes; 80% VLT; Filter number 96
Specifications
Filter Type | 96/Pk. |
Lens Tint | Light Gray |
Product Type | Laser Eyewear |
Transmittance Value | 80% |