North Lightning™ T6550 Series Safety Eyewear
Engineered for safety officers but designed for users
Manufacturer: Honeywell Safety Products T65505B
 View more versions of this product
Catalog No. 19-130-0750
  Check Availability
Add to Cart
    Specifications
    Lens Tint Clear
    Frame Color Black
    Lens Coating 4A Coating (Anti-fog, Anti-scratch, Anti-UV, Anti-static)
    Temple Style Bowed w/soft-touch pads
    Lens Material Polycarbonate
  • Description & Specifications
Related Products
Product Content Feedback