Learn More
Honeywell™ North™ Lightning™ T6550 Series Safety Eyewear
Engineered for safety officers but designed for users
$13.90 - $173.60
Specifications
Lens Type Anti-fog, Anti-static, Anti-UV, Scratch-resistant
Temple Style Bowed w/soft-touch pads
View More Specs
Products 16
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity & Availability  
19-130-0752
View Documents
Honeywell™
T65505BA
Black Amber Pack of 10 for $173.60
Add to cart
19-130-0754
View Documents
Honeywell™
T65505BL
Blue Clear Pack of 10 for $171.24
Add to cart
19-130-0756
View Documents
Honeywell™
T65505BLA
Blue Amber Pack of 10 for $159.90
Add to cart
19-130-0755
View Documents
Honeywell™
T65505BLS
Blue Smoke Pack of 10 for $173.60
Add to cart
19-130-0753
View Documents
Honeywell™
T65505BM
Black Mirror Pack of 10 for $159.00
Add to cart
19-130-0758
View Documents
Honeywell™
T65505S
Silver Silver Pack of 10 for $163.64
Add to cart
19-130-0760
View Documents
Honeywell™
T65505SA
Silver Amber Pack of 10 for $13.90
Add to cart
19-130-0759
View Documents
Honeywell™
T65505SS
Silver Smoke Pack of 10 for $17.36
Add to cart
19-130-0761
View Documents
Honeywell™
T65505SM
Silver Mirror Pack of 10 for $141.00
Add to cart
19-130-0762
View Documents
Honeywell™
T65505BR
Bronze Clear Pack of 10 for $147.32
Add to cart
19-130-0751
View Documents
Honeywell™
T65505BS
Black Smoke Pack of 10 for $14.40
Save 75% Reg : $57.61
Add to cart
19-130-0750
View Documents
Honeywell™
T65505B
Black Clear Pack of 10 for $170.55
Add to cart
19-130-0763
View Documents
Honeywell™
T65505BRS
Bronze Smoke This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-130-0757
View Documents
Honeywell™
T65505BLM
Blue Mirror This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-130-0764
View Documents
Honeywell™
T65505BRA
Bronze Amber This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-130-0765
View Documents
Honeywell™
T65505BRM
Bronze Mirror This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • High base panoramic lens for a wide unobstructed field of view
  • Bowed temples designed to fit virtually anyone
  • New Soft-Touch comfort pads
  • Provided with two Flexi-Fit soft nose-pieces (regular and narrow) for comfort and adaptability
  • The lenses are protected with North's exclusive 4A Coating: Anti-Fog, Anti-Scratch, Anti-UV 99.9%, Anti-Static
  • Meets and/or exceeds ANSI Z87.1 and CSA Z94.3 requirements
Specifications
Specifications
Anti-fog, Anti-static, Anti-UV, Scratch-resistant
Bowed w/soft-touch pads