North™ Low-Profile Cartridges and Cartridge/P100 Filter Combinations

Satisfy numerous industrial applications

Manufacturer: Honeywell Safety Products 75852P100L

 View more versions of this product

Catalog No. 19-003-310B

  Check Availability
Add to Cart

    Specifications

    Quantity 12 Case
    For Use With (Equipment) Respirator,Respirator
    Product Type Mercury Vapor, Chlorine And P100
    For Use With (Application) Industrial applications
    Certifications NIOSH, 42CFR84
    Cartridge Type Cartridge/Filter Assembly
    Color Olive, Magenta

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit