Learn More
Honeywell™ Industrial Weight Rubber Finger Cots
Designed for handling delicate components and subminiature parts
Manufacturer:  Honeywell™ 2315WU/L
 View more versions of this product
Catalog No. 19-069941
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
Cots minimize corrosive effects of perspiration, body oils and salts, hand cream and soap residue. Unrolled.
  • Industrial weight 9 mil cots provide protection for heavy-duty applications
  • Longer wear, better durability than tissue cots
  • Tapered tip is more comfortable and fits fingers better
  • Available in four sizes for better worker fit and comfort
  • Comply with USDA and FDA regulations, 21 CFR, for use in food processing
Specifications
Specifications
Safety
Unrolled
For use in food processing
Large
Finger Cot
White
7 cm
Dipped
For handling delicate components and subminiature parts
Natural Rubber
9mil
No
2.75 in.
ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN374, EN388, EN420, OSHA 29 CFR 1915.157