Honeywell Industrial Weight Rubber Finger Cots

Designed for handling delicate components and subminiature parts

Manufacturer: Honeywell 2115WU/S

 View more versions of this product

Catalog No. 19-070727

  null
Add to Cart

Alert

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Certifications/Compliance ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN374, EN388, EN420, OSHA 29 CFR 1915.157
    Color White
    Construction Dipped
    Cuff Type Unrolled
    For Use With (Application) For use in food processing
    For Use With (Equipment) For handling delicate components and subminiature parts
    Length (English) 2.75 in.
    Length (Metric) 7cm
    Material Natural Rubber
    Powder-Free Yes
    Size Small
    Thickness (English) 9mil
    Product Type Finger Cot

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit