Learn More
Honeywell™ North™ Millennia Sport Safety Glasses
Sleek, lightweight nylon frame with sport design
Manufacturer:  Honeywell™ 11150751
 View more versions of this product
Catalog No. 19-065-144
Add to cart
Description
Description
Sleek, lightweight nylon frame with sport design
Lightweight, comforatble spectacles offer superior side and brow protection with curved temple-to-temple coverage.
  • Superior side and brow protection
  • High impact dual 9-base wraparound polycarbonate unilens
  • Comfortable universal nose bridge
  • Adjustable, breakaway Flexicord™ retainer adds security
Compliance
Meet ANSI™ Z87.1-2003 and CSA Z94.3 standards.
Specifications
Specifications
Meets ANSI Z87.1-2003 and CSA Z94.3 standards.
Black
Polycarbonate
Scratch Resistant
Safety
Hardcoated
TSR Gray