Honeywell North™ Nitri-Knit™ Gloves
Resist cuts, snags, abrasions and punctures
Manufacturer: Honeywell NK803IN/7
 View more versions of this product
Catalog No. 19-150-1051
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR, USDA, FDA
    Glove Cuff Style Trimmed
    For Use With (Application) automotive assembly, painting, chemical handling, processing, degreasing, dip tank cleanup, fishing industry, food processing, general maintenance work, printing cleanup, refinery (general), Solvents (general)
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Lining Insulated
    Material Nitrile
    Size 7
    Coating Material Rough
    Color Blue
  • Description & Specifications
Related Products
Product Content Feedback