Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Anti-Static Nitrile Finger Cots

Powder-free finger cots feature a low particulate count, and help to eliminate damaging static discharge in a wide variety of applications.

$15.83 - $143.70

Specifications

Applications Industrial
Color White
For Use With (Application) Manufacturing Processes
Glove Cuff Style Rolled
Latex Free Yes
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-017-535A
View Documents
Honeywell™ North™
115LNR/S
Small
Pack of 720 for $143.70
Case of 30 PK for $4,103.85
Add to cart
 
19-017-535B
View Documents
Honeywell™ North™
125LNR/MH5
Medium
Pack of 720 for $16.33
Case of 30 PK for $490.02
Add to cart
 
19-017-535C
View Documents
Honeywell™ North™
135LNR/LH5
Large
Pack of 720 for $15.83
Case of 30 PK for $474.83
Add to cart
 
19-017-535D
View Documents
Honeywell™ North™
145LNR/XL
X-Large
Pack of 720 for $134.84
Case of 30 PK for $4,042.71
Edge
Add to cart
 
Description

Description

  • Patented nitrile formulation is comfortable with a high level of stretch
  • Low extractables result in less contamination from reactive ions
  • Helps to eliminate damaging static discharge in a wide variety of applications
  • Free of natural latex proteins that may cause allergic reactions
  • Uniform thickness with excellent tactile sensitivity
Specifications

Specifications

Industrial
Manufacturing Processes
Yes
Nitrile
ESD Finger Cot
White
Rolled
2.8 in.
Powder Free
3 mil
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit