North Nitrile Finger Cots

Powder-Free

Manufacturer: Honeywell Safety Products 115LNR/S

 View more versions of this product

Catalog No. 19-017-535A

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Certifications/Compliance ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN374, EN388, EN420, OSHA 29 CFR 1915.157
    Color White
    Cuff Type Rolled
    For Use With (Application) Eliminates damaging static discharge in many applications and less contamination from reactive ions
    Length (English) 2.75 in.
    Length (Metric) 7cm
    Material Nitrile Rubber
    Powder-Free Yes
    Size Small
    Thickness (English) 3mil
    Product Type Antistatic Finger Cot

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit