North Nitrile Gloves
Manufacturer: Honeywell Safety Products LA172G/10
 View more versions of this product
Catalog No. 17-987-507
  Check Availability
Add to Cart
    Specifications
    Certifications 21 CFR, USDA, FDA
    Glove Cuff Style Straight
    For Use With (Application) aerospace and automotive degreasing, automotive assembling and painting, chemical handling, food processing, graphic arts and print shop cleanup
    Length (English) 13 in.
    Length (Metric) 33cm
    Lining Flock
    Material Nitrile
    Palm Type Resistance to cuts, snags and abrasions
    Size 10
    Thickness 11mil
    Coating Material Sand patch
    Color Green
  • Description & Specifications
Related Products
Product Content Feedback